Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bis(hexyloxy)azoxybenzene 96.0+%, TCI America™
SDP

Catalog No. D10921G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D10921G Supplier TCI America Supplier No. D10921G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2587-42-0
Molecular Formula C24H34N2O3
Molecular Weight (g/mol) 398.547
MDL Number MFCD00043672
InChI Key GWRSINRMEBHRIO-UHFFFAOYSA-N
Synonym 4,4'-dihexyloxyazoxybenzene, 4,4'-bis hexyloxy azoxybenzene, 4,4'-bis n-hexyloxy azoxybenzene, hexoab, 4,4'-dihexoxyazoxybenzene, p,p'-dihexyloxyazoxybenzene, p,p'-bis hexyloxy azoxybenzene, 4,4'-di-n-hexyloxyazoxybenzene, p,p'-bis n-hexyloxy azoxybenzene, azoxybenzene, 4,4'-bis hexyloxy
PubChem CID 75756
IUPAC Name (4-hexoxyphenyl)-(4-hexoxyphenyl)imino-oxidoazanium
SMILES CCCCCCOC1=CC=C(C=C1)N=[N+](C2=CC=C(C=C2)OCCCCCC)[O-]

Specifications

Color Yellow
Quantity 1 g
Formula Weight 398.55
Percent Purity ≥96.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(hexyloxy)azoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.