accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNH2HNHNClNNCl(E/Z)(E)(E)

CAS RN 25875-50-7

IUPAC Name: (E)-N''-[(E)-[(4-chlorophenyl)methylidene]amino]-N-{[(4-chlorophenyl)methylidene]azaniumyl}guanidine chloride
Synonyms: (E)-N''-[(E)-[(4-chlorophenyl)methylidene]amino]-N-{[(4-chlorophenyl)methylidene]ammonio}guanidine chloride
Molecular Weight (g/mol): 370.66
Molecular Formula: C15H14Cl3N5
InChi Key: LTWIBTYLSRDGHP-IQGUDRQUSA-N
SMILES: [Cl-].N\C(N[NH+]=CC1=CC=C(Cl)C=C1)=N/N=C/C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results