Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Robenidine hydrochloride, 98%, Thermo Scientific™

Catalog No. AC459400250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459400250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC459400250 Supplier Thermo Scientific Chemicals Supplier No. 459400250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25875-50-7
Molecular Formula C15H14Cl3N5
Molecular Weight (g/mol) 370.66
MDL Number MFCD01724404
InChI Key LTWIBTYLSRDGHP-IQGUDRQUSA-N
Synonym robenidine hydrochloride, robenidine hcl, unii-8stt15y392, 1,3-bis 4-chlorobenzylidene amino guanidine monohydrochloride, c15h13cl2n5.hcl, 1,3-bis e-4-chlorophenyl methylideneamino guanidine;hydrochloride, chimcoccid hydrochloride, khimcoccid hydrochloride, 1,3-bis 4-chlorobenzylideneamino guanidine hydrochloride
PubChem CID 16212175
IUPAC Name 1,2-bis[(E)-(4-chlorophenyl)methylideneamino]guanidine;hydrochloride
SMILES [Cl-].N\C(N[NH+]=CC1=CC=C(Cl)C=C1)=N/N=C/C1=CC=C(Cl)C=C1

Specifications

Melting Point 252.0°C to 254.0°C
Quantity 25 g
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 370.66
Percent Purity 98%
Chemical Name or Material Robenidine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 247-307-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.