accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3BNNa

CAS RN 25895-62-9

IUPAC Name: sodium cyanoboranuide
Synonyms: sodium cyanoboranuide
Molecular Weight (g/mol): 62.84
Molecular Formula: CH3BNNa
InChi Key: CVDUGUOQTVTBJH-UHFFFAOYSA-N
SMILES: [Na+].[BH3-]C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results