Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Cyanoborodeuteride 98 atom% D, Thermo Scientific™

Catalog No. AC183620010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC183620010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC183620010 Supplier Thermo Scientific Chemicals Supplier No. 183620010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25895-62-9
Molecular Formula CH3BNNa
Molecular Weight (g/mol) 65.86
InChI Key CVDUGUOQTVTBJH-MUTAZJQDSA-N
Synonym sodium cyanoborodeuteride, nabd3cn, sodium;cyano trideuterio boranuide, sodium cyano 2 h? boranuide
PubChem CID 23666334
IUPAC Name sodium;cyano(trideuterio)boranuide
SMILES [BH3-]C#N.[Na+]

Specifications

Physical Form Granular Crystalline Powder
Color White to Yellow
Quantity 1 g
Deuterium (D) 97.5% min.
Linear Formula NaBD3CN
Formula Weight 65.86
Chemical Name or Material Sodium cyanoborodeuteride, 98 atom % D
Hazard Category GHS Signal Word: Danger
Hazard Statement HYGROSCOPIC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.