accessibility menu, dialog, popup

UserName

Viewing profile as user:
ON

CAS RN 2591-86-8

IUPAC Name: piperidine-1-carbaldehyde
Synonyms: N-formylpiperidine
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: FEWLNYSYJNLUOO-UHFFFAOYSA-N
SMILES: O=CN1CCCCC1
Boiling Point 
  • (8)
  • (2)
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (8)
  • (2)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)