Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Formylpiperidine

Catalog No. F17407500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F17407500G 500 g
F17407100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F17407500G Supplier Sigma Aldrich Supplier No. F17407500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reactant for:
  • Direct amidation of azoles with formamides via metal-free c-h activation in the presence of tert-butyl perbenzoate
  • Vilsmeier-type reactions of pyrazoles with amides in the presence of phosphorous oxychloride
  • Co-free aminocarbonylation of n-substituted formamides with aryl iodides/bromides catalyzed by palladium acetate and xantphos for synthesis of arylamides
  • Synthesis of alternating copolymers for organic photovoltaic applications
  • One-pot double functionalization of -deficient heterocyclic lithium reagents
  • Synthesis of thermally stable piezofluorochromic aggregation-induced emission compounds

Specifications

CAS 2591-86-8
Density 1.019 g/mL (at 25°C (literature))
Boiling Point 222°C (lit.)
Molecular Formula C6H11NO
Refractive Index n20/D 1.484 (literature)
Linear Formula C6H11NO
MDL Number MFCD00006483
Quantity 500 g
Synonym N-Carbaldehyde piperidine; NFP; Piperidine-1-carboxaldehyde
Molecular Weight (g/mol) 113.16
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.