accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOONCH3H3COSOOO(E/Z)

CAS RN 25910-85-4

IUPAC Name: 6-ethoxy-1-methyl-2-[2-(3-nitrophenyl)ethenyl]quinolin-1-ium methyl sulfate
Synonyms: 6-ethoxy-1-methyl-2-[2-(3-nitrophenyl)ethenyl]quinolin-1-ium methyl sulfate(1-)
Molecular Weight (g/mol): 446.47
Molecular Formula: C21H22N2O7S
InChi Key: ZXQHSPWBYMLHLB-UHFFFAOYSA-M
SMILES: COS([O-])(=O)=O.CCOC1=CC=C2C(C=CC(C=CC3=CC=CC(=C3)[N+]([O-])=O)=[N+]2C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results