Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pinacryptol Yellow, MP Biomedicals

Catalog No. ICN216858 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN216858 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN216858 Supplier MP Biomedicals, Inc Supplier No. 0521685801

Chemical Identifiers

CAS 25910-85-4
Molecular Formula C21H22N2O7S
Molecular Weight (g/mol) 446.47
MDL Number MFCD00011967
InChI Key ZXQHSPWBYMLHLB-UHFFFAOYSA-M
PubChem CID 6438045
IUPAC Name 6-ethoxy-1-methyl-2-[2-(3-nitrophenyl)ethenyl]quinolin-1-ium methyl sulfate
SMILES COS([O-])(=O)=O.CCOC1=CC=C2C(C=CC(C=CC3=CC=CC(=C3)[N+]([O-])=O)=[N+]2C)=C1

Specifications

Formula Weight 446.48
Quantity 1 g
Chemical Name or Material Pinacryptol Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.