accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 259269-84-6

IUPAC Name: 2-amino-3-bromo-5-fluorobenzoic acid
Synonyms: 2-amino-3-bromo-5-fluorobenzoic acid
Molecular Weight (g/mol): 234.02
Molecular Formula: C7H5BrFNO2
InChi Key: LTOCPZGBNWZMBA-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(F)C=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results