Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-bromo-5-fluorobenzoic acid, 95%, Thermo Scientific™

Catalog No. AAH66628MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66628MD 250 mg
AAH6662803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66628MD Supplier Thermo Scientific Chemicals Supplier No. H66628MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 259269-84-6
Molecular Formula C7H5BrFNO2
Molecular Weight (g/mol) 234.024
MDL Number MFCD00662509
InChI Key LTOCPZGBNWZMBA-UHFFFAOYSA-N
Synonym benzoic acid, 2-amino-3-bromo-5-fluoro, 2-amino-3-bromo-5-fluoro-benzoic acid, 2-amino-3-bromo-5-fluorobenzoicacid
PubChem CID 2781703
IUPAC Name 2-amino-3-bromo-5-fluorobenzoic acid
SMILES C1=C(C=C(C(=C1Br)N)C(=O)O)F

Specifications

Quantity 250 mg
Beilstein 15583945
Formula Weight 234.02
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-3-bromo-5-fluorobenzoic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.