accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)OOH(R)OH(R)HO(S)HO

CAS RN 2595-97-3

IUPAC Name: (3R,4R,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Synonyms: D-allopyranose D-allose
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: WQZGKKKJIJFFOK-IVMDWMLBSA-N
SMILES: OC[C@H]1OC(O)[C@H](O)[C@H](O)[C@@H]1O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)