accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaH3COCH3NHNOSOOOSOOO(E)

CAS RN 25956-17-6

IUPAC Name: disodium (5E)-5-[2-(2-methoxy-5-methyl-4-sulfonatophenyl)hydrazin-1-ylidene]-6-oxo-5,6-dihydronaphthalene-2-sulfonate
Synonyms: curry red
Molecular Weight (g/mol): 496.42
Molecular Formula: C18H14N2Na2O8S2
InChi Key: POXPUHKJGLHZEM-VIPPSAFOSA-L
SMILES: [Na+].[Na+].COC1=CC(=C(C)C=C1N\N=C1\C(=O)C=CC2=CC(=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (5)
  • (1)
  • (1)
Grade 
  • (1)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (5)