Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Red Dye 0.5% Solution, Innovating Science™

Catalog No. S001298 Shop All Aldon Products
Change view
Click to view available options
Quantity:
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001298 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001298 Supplier Aldon Corporation Supplier No. IS35061
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-quality dye in a 0.5% aqueous solution, suitable for laboratory activity kits and science applications.

  • 30 mL aqueous solution.
  • Ideal for laboratory activity kits focusing on chromatography spectroscopy or dilution experiments.
  • A consistent and reliable resource for hands-on science and research use.

Specifications

Chemical Name or Material Red #40
CAS 25956-17-6, 7732-18-5
Color Red
Quantity 30 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.