accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOHO(E/Z)(Z)

CAS RN 25959-50-6

IUPAC Name: (2Z)-3-(4-{[(4-methoxyphenyl)methylidene]amino}phenyl)prop-2-enoic acid
Synonyms: (2Z)-3-(4-{[(4-methoxyphenyl)methylidene]amino}phenyl)prop-2-enoic acid
Molecular Weight (g/mol): 281.31
Molecular Formula: C17H15NO3
InChi Key: UIELBEHBZKVMEI-MGGYQKONSA-N
SMILES: COC1=CC=C(C=NC2=CC=C(\C=C/C(O)=O)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results