Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[(4-Methoxybenzylidene)amino]cinnamic Acid 98.0+%, TCI America™
SDP

Catalog No. M05881G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M05881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M05881G Supplier TCI America Supplier No. M05881G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25959-50-6
Molecular Formula C17H15NO3
Molecular Weight (g/mol) 281.311
MDL Number MFCD00021777
InChI Key UIELBEHBZKVMEI-MGGYQKONSA-N
Synonym 4-(p-Anisalamino)cinnamic Acid, 4-[(p-Anisylidene)amino]cinnamic Acid
PubChem CID 5357579
IUPAC Name (Z)-3-[4-[(4-methoxyphenyl)methylideneamino]phenyl]prop-2-enoic acid
SMILES COC1=CC=C(C=C1)C=NC2=CC=C(C=C2)C=CC(=O)O

Specifications

Melting Point 198°C
Color Yellow
Quantity 1 g
Formula Weight 281.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[(4-Methoxybenzylidene)amino]cinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.