accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH2NSH

CAS RN 25984-63-8

IUPAC Name: 4-[amino(sulfanyl)methylidene]cyclohexa-2,5-dien-1-one
Synonyms: 4-[amino(sulfanyl)methylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 153.2
Molecular Formula: C7H7NOS
InChi Key: DOCQBKMMNPJLOR-UHFFFAOYSA-N
SMILES: NC(S)=C1C=CC(=O)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)