accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25990-96-9

IUPAC Name: 2,5,8,11,14,17,20,23-octaoxapentacosan-25-ol
Synonyms: 2,5,8,11,14,17,20,23-octaoxapentacosan-25-ol
Molecular Weight (g/mol): 384.47
Molecular Formula: C17H36O9
InChi Key: SZGNWRSFHADOMY-UHFFFAOYSA-N
SMILES: COCCOCCOCCOCCOCCOCCOCCOCCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results