accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 260-50-4

IUPAC Name: benzo[g]quinoxaline
Synonyms: benzo[g]quinoxaline
Molecular Weight (g/mol): 180.21
Molecular Formula: C12H8N2
InChi Key: XEMRLVBSKVCUDL-UHFFFAOYSA-N
SMILES: C1=CC=C2C=C3N=CC=NC3=CC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results