Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[g]quinoxaline 98.0+%, TCI America™
SDP

Catalog No. B4540200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4540200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4540200MG Supplier TCI America Supplier No. B4540200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 260-50-4
Molecular Formula C12H8N2
Molecular Weight (g/mol) 180.21
MDL Number MFCD18449114
InChI Key XEMRLVBSKVCUDL-UHFFFAOYSA-N
PubChem CID 12886698
IUPAC Name benzo[g]quinoxaline
SMILES C1=CC=C2C=C3N=CC=NC3=CC2=C1

Specifications

Melting Point 127°C
Color Yellow
Quantity 200 mg
Formula Weight 180.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzo[g]quinoxaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.