accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2602-46-2

IUPAC Name: tetrasodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-8-amino-1-oxo-3,6-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: tetrasodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-8-amino-1-oxo-3,6-disulfonatonaphthalen-2-ylidene]hydrazin-1-yl}-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxonaphthalene-2,7-disulfonate
Molecular Weight (g/mol): 932.74
Molecular Formula: C32H20N6Na4O14S4
InChi Key: HOPRIFATEWRIEO-YSKRITFZSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].NC1=CC(=CC2=C1C(=O)\C(=N\NC1=CC=C(C=C1)C1=CC=C(N\N=C3\C(=O)C4=C(N)C=C(C=C4C=C3S([O-])(=O)=O)S([O-])(=O)=O)C=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results