Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Direct Blue 6, TCI America™
SDP

Catalog No. D02351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D02351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. D02351G Supplier TCI America Supplier No. D02351G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 2602-46-2
Molecular Formula C32H20N6Na4O14S4
Molecular Weight (g/mol) 932.739
MDL Number MFCD00068426
InChI Key HOPRIFATEWRIEO-UHFFFAOYSA-J
Synonym Benzo Blue 2B, Nippon Blue B
PubChem CID 73557447
IUPAC Name tetrasodium;5-amino-3-[[4-[4-[2-(8-amino-1-oxo-3,6-disulfonatonaphthalen-2-ylidene)hydrazinyl]phenyl]phenyl]hydrazinylidene]-4-oxonaphthalene-2,7-disulfonate
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)NN=C3C(=CC4=CC(=CC(=C4C3=O)N)S(=O)(=O)[O-])S(=O)(=O)[O-])NN=C5C(=CC6=CC(=CC(=C6C5=O)N)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Color Black-Gray
Quantity 1 g
Formula Weight 932.74
Physical Form Crystalline Powder
Chemical Name or Material Direct Blue 6
EINECSNumber (5)-1283
RTECSNumber QJ6400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.