accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHN

CAS RN 26033-20-5

IUPAC Name: 5-phenyl-1H-pyrazole-4-carbaldehyde
Synonyms: 3-phenyl-2H-pyrazole-4-carbaldehyde
Molecular Weight (g/mol): 172.19
Molecular Formula: C10H8N2O
InChi Key: OCCFXKQCKSLEII-UHFFFAOYSA-N
SMILES: O=CC1=C(NN=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results