Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenyl-1H-pyrazole-4-carboxaldehyde, 98%, Thermo Scientific™

Catalog No. AC296000010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296000010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC296000010 Supplier Thermo Scientific Chemicals Supplier No. 296000010

Chemical Identifiers

CAS 26033-20-5
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.19
MDL Number MFCD00216971
InChI Key OCCFXKQCKSLEII-UHFFFAOYSA-N
Synonym 3-phenyl-1h-pyrazole-4-carbaldehyde, 3-phenyl-1h-pyrazole-4-carboxaldehyde, 3-phenylpyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 3-phenyl, 3-phenyl-2h-pyrazole-4-carbaldehyde, 5-phenylpyrazole-4-carbaldehyde, pubchem16274, acmc-1cd6r, 3-phenyl-4-formylpyrazole, cambridge id 5132647
PubChem CID 291816
IUPAC Name 5-phenyl-1H-pyrazole-4-carbaldehyde
SMILES O=CC1=C(NN=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 146°C to 148°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: 25mg/mL (100°C). Other solubilities: 400mg/mL ethanol (78°C)
Formula Weight 172.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Phenyl-1H-pyrazole-4-carboxaldehyde, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.