accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2605-14-3

IUPAC Name: 2-chloro-6-methoxy-1,3-benzothiazole
Synonyms: 2-chloro-6-methoxy-1,3-benzothiazole
Molecular Weight (g/mol): 199.65
Molecular Formula: C8H6ClNOS
InChi Key: FVUFTABOJFRHSU-UHFFFAOYSA-N
SMILES: COC1=CC=C2N=C(Cl)SC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results