CAS RN 2609-88-3
CAS RN 2609-88-3
IUPAC Name:
sodium 3,6-bis(diethylamino)-9-(2,4-disulfonatophenyl)-2,7-dimethyl-10λ⁴-xanthen-10-ylium
Synonyms:
sulforhodamine B acid rhodamine B amido rhodamine B xylene red B acid red XB acid red 52 food red 106 kiton rhodamine B lissamine rhodamine
Molecular Weight (g/mol):
608.7
Molecular Formula:
C29H33N2NaO7S2
InChi Key:
XJENLUNLXRJLEZ-UHFFFAOYSA-M
SMILES:
[Na+].CCN(CC)C1=CC2=[O+]C3=CC(N(CC)CC)=C(C)C=C3C(C3=CC=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C2C=C1C
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich Sulforhodamine B, acid form
Sigma Aldrich Sulforhodamine B, acid form
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.