accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2610-11-9

NaNaOSOONNNHNONHOSOOO(E/Z)(E)

CAS RN 2610-11-9

IUPAC Name: disodium (3E)-7-benzamido-4-oxo-3-(2-{4-[2-(4-sulfonatophenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-3,4-dihydronaphthalene-2-sulfonate
Synonyms: disodium (3E)-7-benzamido-4-oxo-3-(2-{4-[2-(4-sulfonatophenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)naphthalene-2-sulfonate
Molecular Weight (g/mol): 675.6
Molecular Formula: C29H19N5Na2O8S2
InChi Key: OTPBDPKSNGORHR-ATQPHANLSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(N\N=C2/C(=O)C3=CC=C(NC(=O)C4=CC=CC=C4)C=C3C=C2S([O-])(=O)=O)C=C1
Color 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results