Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Direct Red 81, Sigma-Aldrich

Catalog No. 01258852 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-852 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-852 Supplier MilliporeSigma Supelco Supplier No. 195251100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bacterial consortium NBNJ6 has an ability to decolorize direct red 81 (DR81).

TRUSTED_SUSTAINABILITY

Specifications

CAS 2610-11-9
Molecular Weight (g/mol) 675.6 g/mol
Color Dark Red
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.