accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOHO

CAS RN 26100-51-6

IUPAC Name: 2-hydroxypropanoic acid
Synonyms: lactic acid milchsaeure
Molecular Weight (g/mol): 90.08
Molecular Formula: C3H6O3
InChi Key: JVTAAEKCZFNVCJ-UHFFFAOYNA-N
SMILES: CC(O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results