accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26112-50-5

IUPAC Name: (2-hydroxypropyl)trimethylazanium iodide
Synonyms: β-methylcholine iodide
Molecular Weight (g/mol): 245.1
Molecular Formula: C6H16INO
InChi Key: FOILINVEQJRMPF-UHFFFAOYNA-M
SMILES: [I-].CC(O)C[N+](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results