Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Methylcholine Iodide 99.0+%, TCI America™
SDP

Catalog No. M059825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M059825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M059825G Supplier TCI America Supplier No. M059825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26112-50-5
Molecular Formula C6H16INO
Molecular Weight (g/mol) 245.10
MDL Number MFCD00059987
InChI Key FOILINVEQJRMPF-UHFFFAOYNA-M
Synonym 2-Hydroxypropyltrimethylammonium Iodide
PubChem CID 10263892
IUPAC Name (2-hydroxypropyl)trimethylazanium iodide
SMILES [I-].CC(O)C[N+](C)(C)C

Specifications

Melting Point 160°C
Color White
Quantity 25 g
Formula Weight 245.10
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material beta-Methylcholine Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.