accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHHOO

CAS RN 2612-02-4

IUPAC Name: 2-hydroxy-5-methoxybenzoic acid
Synonyms: 5-methoxysalicylic acid acid5-methoxysalicylic
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: IZZIWIAOVZOBLF-UHFFFAOYSA-N
SMILES: COC1=CC=C(O)C(=C1)C(O)=O
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)