accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2613-23-2

IUPAC Name: 3-chloro-4-fluorophenol
Synonyms: 3-chloro-4-fluorophenol
Molecular Weight (g/mol): 146.55
Molecular Formula: C6H4ClFO
InChi Key: ZQXLIXHVJVAPLW-UHFFFAOYSA-N
SMILES: OC1=CC=C(F)C(Cl)=C1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)