Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-fluorophenol 96.0+%, TCI America™
SDP

Catalog No. C112110G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C112110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C112110G Supplier TCI America Supplier No. C112110G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2613-23-2
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.545
MDL Number MFCD00002257
InChI Key ZQXLIXHVJVAPLW-UHFFFAOYSA-N
Synonym phenol, 3-chloro-4-fluoro, 3-chloro-4-fluoro-phenol, pubchem1492, pubchem2856, 3-chloro-4-fluorophe, 3-chloro4-fluorophenol, acmc-209goi, 4-fluoro-3-chlorophenol, 3-chloro-4-fluorophenol, ksc204s3j
PubChem CID 75790
IUPAC Name 3-chloro-4-fluorophenol
SMILES C1=CC(=C(C=C1O)Cl)F

Specifications

Melting Point 40°C
Color White-Yellow
Boiling Point 104°C
Quantity 10 g
Formula Weight 146.55
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Chloro-4-fluorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.