accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONH(R)BrOO

CAS RN 261360-76-3

IUPAC Name: (2R)-3-(2-bromophenyl)-2-{[(tert-butoxy)carbonyl]amino}propanoate
Synonyms: (2R)-3-(2-bromophenyl)-2-[(tert-butoxycarbonyl)amino]propanoate
Molecular Weight (g/mol): 343.2
Molecular Formula: C14H17BrNO4
InChi Key: XDJSTMCSOXSTGZ-LLVKDONJSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1Br)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)