Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-2-bromo-D-phenylalanine, 95%, Thermo Scientific™

Catalog No. AAH62848MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62848MD 250 mg
AAH6284803 1 g
AAH6284806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH62848MD Supplier Thermo Scientific Chemicals Supplier No. H62848MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Boc-2-bromo-D-phenylalanine is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261360-76-3
Molecular Formula C14H18BrNO4
Molecular Weight (g/mol) 344.205
MDL Number MFCD01317703
InChI Key XDJSTMCSOXSTGZ-LLVKDONJSA-N
Synonym boc-2-bromo-d-phenylalanine, boc-d-phe 2-br-oh, n-boc-2-bromo-d-phenylalanine, boc-d-2-bromophenylalanine, r-n-boc-2-bromophenylalanine, boc-d-2-bromophe, r-3-2-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-2-br-phe-oh, 2r-3-2-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, n-boc-d-2-bromophenylalanine
PubChem CID 7006627
IUPAC Name (2R)-3-(2-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1Br)C(=O)O

Specifications

Color White
Quantity 250 mg
Formula Weight 344.2
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-2-bromo-D-phenylalanine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.