accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2615-15-8

IUPAC Name: 3,6,9,12,15-pentaoxaheptadecane-1,17-diol
Synonyms: polyethylene glycol 300 polyoxyethylene (6) hexaoxyethylene glycol hexagol
Molecular Weight (g/mol): 282.33
Molecular Formula: C12H26O7
InChi Key: IIRDTKBZINWQAW-UHFFFAOYSA-N
SMILES: OCCOCCOCCOCCOCCOCCO
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)