accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOSiCH3H3CCH3CH3H3CHOOH

CAS RN 261621-12-9

IUPAC Name: {3-[(tert-butyldimethylsilyl)oxy]phenyl}boronic acid
Synonyms: 3-[(tert-butyldimethylsilyl)oxy]phenylboronic acid
Molecular Weight (g/mol): 252.19
Molecular Formula: C12H21BO3Si
InChi Key: RDQWADDNQONTLB-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OC1=CC=CC(=C1)B(O)O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)