Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(tert-Butyldimethylsilyloxy)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B37265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3726-5G 5 g
B37261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3726-5G Supplier TCI America Supplier No. B37265G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 261621-12-9
Melting Point 91°C
Color White-Yellow
Molecular Formula C12H21BO3Si
Quantity 5 g
MDL Number MFCD03701505
Synonym 3-tert-butyldimethylsilyl oxy phenyl boronic acid, 3-tert-butyldimethylsilyloxy phenylboronic acid, 3-t-butyldimethylsilyloxy phenylboronic acid, 3-tert-butyl dimethylsiloxy phenyl boronic acid, 3-tert-butyldimethylsilyloxy benzeneboronic acid
InChI Key RDQWADDNQONTLB-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)O[Si](C)(C)C(C)(C)C)(O)O
IUPAC Name [3-[tert-butyl(dimethyl)silyl]oxyphenyl]boronic acid
Molecular Weight (g/mol) 252.192
PubChem CID 3463029
Formula Weight 252.19
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.