accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26165-45-7

IUPAC Name: 3-(pyrrolidin-1-yl)propanenitrile
Synonyms: 1-pyrrolidinepropionitrile
Molecular Weight (g/mol): 124.19
Molecular Formula: C7H12N2
InChi Key: PDVYZQGNSCSKPG-UHFFFAOYSA-N
SMILES: N#CCCN1CCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)