Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pyrrolidinepropionitrile, 97%, Thermo Scientific™

Catalog No. AAL0181814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0181814 25 g
AAL0181806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0181814 Supplier Thermo Scientific Chemicals Supplier No. L0181814

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26165-45-7
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.187
MDL Number MFCD00014099
InChI Key PDVYZQGNSCSKPG-UHFFFAOYSA-N
Synonym 1-pyrrolidinepropionitrile, 3-pyrrolidin-1-yl propanenitrile, 3-1-pyrrolidino propionitrile, 1-pyrrolidinepropanenitrile, beta-pyrrolidinopropionitrile, n-2-cyanoethyl pyrrolidine, pyrrolidine-1-propiononitrile, n-beta-cyanoethyl pyrrolidine, 3-1-pyrrolidine propionitrile, 3-1-pyrrolidinyl propionitrile
PubChem CID 117743
IUPAC Name 3-pyrrolidin-1-ylpropanenitrile
SMILES C1CCN(C1)CCC#N

Specifications

Boiling Point 104°C to 105°C (15 mmHg)
Refractive Index 1.466
Quantity 25 g
UN Number UN3276
Beilstein 107542
Formula Weight 124.19
Percent Purity 97%
Chemical Name or Material 1-Pyrrolidinepropionitrile
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 247-493-0
RTECSNumber UY2273000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.