accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNO

CAS RN 261715-39-3

IUPAC Name: 6-(pyrrolidin-1-yl)pyridine-3-carbaldehyde
Synonyms: 6-(pyrrolidin-1-yl)pyridine-3-carbaldehyde
Molecular Weight (g/mol): 176.22
Molecular Formula: C10H12N2O
InChi Key: FFFAXGTZBUMKTI-UHFFFAOYSA-N
SMILES: O=CC1=CN=C(C=C1)N1CCCC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)