Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(1-Pyrrolidinyl)nicotinaldehyde, Thermo Scientific™

Catalog No. MO07079CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO07079CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07079CB Supplier Thermo Scientific Chemicals Supplier No. MO07079CB
Add to Cart

Chemical Identifiers

CAS 261715-39-3
Molecular Formula C10H12N2O
Molecular Weight (g/mol) 176.219
InChI Key FFFAXGTZBUMKTI-UHFFFAOYSA-N
Synonym 6-1-pyrrolidinyl nicotinaldehyde, 6-pyrrolidin-1-yl nicotinaldehyde, 6-pyrrolidin-1-yl pyridine-3-carbaldehyde, 6-pyrrolidin-1-yl pyridine-3-carboxaldehyde, 6-1-pyrrolidinyl pyridine-3-carboxaldehyde, 6-pyrrolidin-1-ylnicotinaldehyde, 6-pyrrolidinylpyridine-3-carbaldehyde, 6-1-pyrrolidino pyridine-3-carboxaldehyde, acmc-1cg5q, 6-1-pyrrolidino nicotinaldehyde
PubChem CID 2794775
IUPAC Name 6-pyrrolidin-1-ylpyridine-3-carbaldehyde
SMILES C1CCN(C1)C2=NC=C(C=C2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 176.22
Percent Purity 97%
Chemical Name or Material 6-(1-Pyrrolidinyl)nicotinaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.