accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3NSO

CAS RN 26172-54-3

IUPAC Name: hydrogen 2-methyl-2,3-dihydro-1,2-thiazol-3-one chloride
Synonyms: methylisothiazolinone
Molecular Weight (g/mol): 151.61
Molecular Formula: C4H6ClNOS
InChi Key: SJXPQSRCFCPWQQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN1SC=CC1=O
Color 
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)