accessibility menu, dialog, popup

UserName

Viewing profile as user:
PONHPPhPhPhPh

CAS RN 261733-18-0

IUPAC Name: 4,6-bis(diphenylphosphanyl)-10H-phenoxazine
Synonyms: 4,6-bis(diphenylphosphanyl)-10H-phenoxazine
Molecular Weight (g/mol): 551.57
Molecular Formula: C36H27NOP2
InChi Key: HSWZLYXRAOXOLL-UHFFFAOYSA-N
SMILES: N1C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)