Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Bis(diphenylphosphino)phenoxazine 98.0+%, TCI America™
SDP

Catalog No. B27171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27171G 1 g
B2717100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27171G Supplier TCI America Supplier No. B27171G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 261733-18-0
Molecular Formula C36H27NOP2
Molecular Weight (g/mol) 551.57
MDL Number MFCD03788937
InChI Key HSWZLYXRAOXOLL-UHFFFAOYSA-N
Synonym 4,6-bis diphenylphosphino phenoxazine, 4,6-bis diphenylphosphino-10h-phenoxazine, nixantphos, 10h-phenoxazine, 4,6-bis diphenylphosphino, 4,6-bis diphenylphosphanyl-10h-phenoxazine, nixanthphos, n-xantphos, acmc-209gp1, bis diphenylphosphino phenoxazine, 4,6-bis diphenylphosphino-phenoxazine
PubChem CID 3255346
IUPAC Name 4,6-bis(diphenylphosphanyl)-10H-phenoxazine
SMILES N1C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 551.57
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Bis(diphenylphosphino)phenoxazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.