accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFClF

CAS RN 261762-51-0

IUPAC Name: 3-chloro-2,6-difluorophenol
Synonyms: 3-chloro-2,6-difluorophenol
Molecular Weight (g/mol): 164.54
Molecular Formula: C6H3ClF2O
InChi Key: CCLYWHXHYLQWQK-UHFFFAOYSA-N
SMILES: OC1=C(F)C=CC(Cl)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results