accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26177-43-5

IUPAC Name: hydrogen 1-(3-nitrophenyl)methanamine chloride
Synonyms: hydrogen 1-(3-nitrophenyl)methanamine chloride
Molecular Weight (g/mol): 188.61
Molecular Formula: C7H9ClN2O2
InChi Key: DLZXLCHQWOZGSE-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC1=CC=CC(=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results