accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSOHNSOO

CAS RN 2618-96-4

IUPAC Name: N-(benzenesulfonyl)benzenesulfonamide
Synonyms: N-(benzenesulfonyl)benzenesulfonamide
Molecular Weight (g/mol): 297.34
Molecular Formula: C12H11NO4S2
InChi Key: OVQABVAKPIYHIG-UHFFFAOYSA-N
SMILES: O=S(=O)(NS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)