Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzenesulfonamide, 95%

Catalog No. AAH60569MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60569MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH60569MD Supplier Thermo Scientific Chemicals Supplier No. H60569MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2618-96-4 | C12H11NO4S2 | 297.343 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2618-96-4
Molecular Formula C12H11NO4S2
Molecular Weight (g/mol) 297.343
MDL Number MFCD00025019
InChI Key OVQABVAKPIYHIG-UHFFFAOYSA-N
Synonym dibenzenesulfonimide, benzenesulfonamide, n-phenylsulfonyl, dibenzenesulfonamide, bis phenylsulfonyl amine, n-phenylsulfonyl benzenesulfonamide, dibenzolsulfimid, dibenzolsulfimide, diphenylsulfonimide, benzenesulfonic acid imide, n-benzenesulfonyl benzenesulfonamide
PubChem CID 75671
IUPAC Name N-(benzenesulfonyl)benzenesulfonamide
SMILES C1=CC=C(C=C1)S(=O)(=O)NS(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 150°C
Quantity 250 mg
Formula Weight 297.35
Percent Purity 95%
Chemical Name or Material Dibenzenesulfonamide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 220-051-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.